Skip to Content
Merck

Skip To

NMIM296

dl-Norephedrine (Phenylpropanolamine) hydrochloride

NMI Australia

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C9H14ClNO
CAS Number:
Molecular Weight:
187.67
UNSPSC Code:
41116107
EC Number:
205-826-7


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

room temp

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9-;/m1./s1

InChI key

DYWNLSQWJMTVGJ-PRCZDLBKSA-N



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters