Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H15NO2 · HCl
CAS Number:
Molecular Weight:
217.69
UNSPSC Code:
41116107
grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
room temp
InChI
1S/C10H15NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,10-13H,2,7H2,1H3;1H
InChI key
KTNROWWHOBZQGK-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Etilefrine hydrochloride
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 3
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
