Skip to Content
Merck

Skip To

NMIS008

Boldenone (1,4-androstdien-17β-ol-3-one)

NMI Australia


About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
UNSPSC Code:
41116107
EC Number:
212-686-0
eCl@ss:
25140904


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

2-8°C

InChI

1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h7,9,11,14-17,21H,3-6,8,10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1

InChI key

RSIHSRDYCUFFLA-DYKIIFRCSA-N



Pictograms

Health hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters