Skip to Content
Merck

Skip To

NMIS021

Methasterone

NMI Australia

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
UNSPSC Code:
41116107


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

2-8°C

InChI

1S/C21H34O2/c1-13-12-19(2)14(11-18(13)22)5-6-15-16(19)7-9-20(3)17(15)8-10-21(20,4)23/h13-17,23H,5-12H2,1-4H3/t13-,14+,15-,16+,17+,19+,20+,21+/m1/s1

InChI key

QCWCXSMWLJFBNM-FOVYBZIDSA-N



pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters