About This Item
Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
2-8°C
InChI
1S/C27H44O3/c1-5-6-7-8-9-25(29)30-24-13-12-21-20-11-10-19-16-23(28)18(2)17-27(19,4)22(20)14-15-26(21,24)3/h18-22,24H,5-17H2,1-4H3/t18-,19+,20?,21?,22?,24+,26+,27+/m1/s1
InChI key
ZNYZYASFVYLKPE-SRGKWKMDSA-N
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
