Skip to Content
Merck

Skip To

NMIS039

3alpha-5-Cyclo-5alpha-androstane-6beta-ol-17-one

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
UNSPSC Code:
41116107


grade

certified reference material

agency

NMI Australia certified reference material

form

solid

manufacturer/tradename

NMI Australia

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

2-8°C

InChI

1S/C19H28O2/c1-17-7-6-14-12(13(17)3-4-15(17)20)9-16(21)19-10-11(19)5-8-18(14,19)2/h11-14,16,21H,3-10H2,1-2H3/t11-,12-,13-,14-,16+,17-,18+,19-/m0/s1

InChI key

NIDVKCKYZAQGOI-LYVQZGCSSA-N



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters