Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C19H30O3
CAS Number:
Molecular Weight:
306.44
UNSPSC Code:
41116107
eCl@ss:
32150313
grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
2-8°C
InChI
1S/C19H30O3/c1-17-11-22-16(20)10-12(17)4-5-13-14(17)6-8-18(2)15(13)7-9-19(18,3)21/h12-15,21H,4-11H2,1-3H3/t12-,13+,14-,15-,17-,18-,19-/m0/s1
InChI key
QSLJIVKCVHQPLV-PEMPUTJUSA-N
Still not finding the right product?
Explore all of our products under Oxandrolone
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
