Skip to Content
Merck

COM448616993

Combi-Blocks, Inc.

N-Cyclopropyl 4-bromo-3-methoxybenzamide

Synonym(s):

4-bromo-N-cyclopropyl-3-methoxybenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H12O2N1Br1
CAS Number:
Molecular Weight:
270.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H12BrNO2/c1-15-10-6-7(2-5-9(10)12)11(14)13-8-3-4-8/h2,5-6,8H,3-4H2,1H3,(H,13,14)

Inchi Key

ACZUAJCGIAVKNK-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library