Skip to Content
Merck

COM448638643

Combi-Blocks, Inc.

Minoxidil sulfate

Synonym(s):

2,6-diamino-4-(1-piperidinyl)-1-(sulfonatooxy)pyrimidin-1-ium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H15O4N5S1
CAS Number:
Molecular Weight:
289.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H15N5O4S/c10-7-6-8(13-4-2-1-3-5-13)12-9(11)14(7)18-19(15,16)17/h6H,1-5H2,(H4,10,11,12,15,16,17)

Inchi Key

OEOLOEUAGSPDLT-UHFFFAOYSA-N

Purity

98%

Storage temp.

Freezer

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library