Inchi Code
1S/C12H14N2O3S.Na.H2O/c13-7-8-14-11-5-1-4-10-9(11)3-2-6-12(10)18(15,16)17;;/h1-6,14H,7-8,13H2,(H,15,16,17);;1H2/q;+1;/p-1
Inchi Key
RSSNFBRKWCVQDM-UHFFFAOYSA-M
Purity
95%
Country of Origin
CN
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.