Skip to Content
Merck

COMH93D5CC9D

Combi-Blocks, Inc.

N-(5-Chloro-2-hydroxyphenyl)acetamide

Synonym(s):

N-(5-chloro-2-hydroxyphenyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O2N1Cl1
CAS Number:
Molecular Weight:
185.61
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H8ClNO2/c1-5(11)10-7-4-6(9)2-3-8(7)12/h2-4,12H,1H3,(H,10,11)

Inchi Key

HGDAFIJKXCFHPG-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library