Skip to Content
Merck

COMH95E96CE4

Combi-Blocks, Inc.

Imazapyr

Synonym(s):

2-(4-isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)nicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H15O3N3
CAS Number:
Molecular Weight:
261.28
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H15N3O3/c1-7(2)13(3)12(19)15-10(16-13)9-8(11(17)18)5-4-6-14-9/h4-7H,1-3H3,(H,17,18)(H,15,16,19)

Inchi Key

CLQMBPJKHLGMQK-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



Still not finding the right product?

Explore all of our products under Imazapyr




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library