Skip to Content
Merck

Skip To

COMH96ED8167

Combi-Blocks, Inc.

TE buffer

Synonym(s):

2-amino-2-(hydroxymethyl)propane-1,3-diol 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C14H27O11N3
CAS Number:
Molecular Weight:
413.38
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H16N2O8.C4H11NO3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2

Inchi Key

VLEIUWBSEKKKFX-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library