Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C6H2Cl2O4.2Na/c7-1-3(9)5(11)2(8)6(12)4(1)10;;/h9,12H;;/q;2*+1/p-2
Inchi Key
FITANBHHCLWRKG-UHFFFAOYSA-L
Purity
95%
Country of Origin
CN
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.