Skip to Content
Merck

COMH97F0DEC8

Combi-Blocks, Inc.

2,6-Dibromo-4-isopropylphenol

Synonym(s):

2,6-dibromo-4-isopropylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O1Br2
Molecular Weight:
293.99
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10Br2O/c1-5(2)6-3-7(10)9(12)8(11)4-6/h3-5,12H,1-2H3

Inchi Key

DOSJZFDHMKBPRM-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library