Skip to Content
Merck

COMH99E24366

Combi-Blocks, Inc.

2-Bromo-6-fluorobiphenyl

Synonym(s):

2-bromo-6-fluoro-1,1'-biphenyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H8Br1F1
Molecular Weight:
251.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H8BrF/c13-10-7-4-8-11(14)12(10)9-5-2-1-3-6-9/h1-8H

Inchi Key

CJRQBSHIUWBRKE-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library