Skip to Content

MATH03871FCA

Matrix Scientific

2-(Bromomethyl)pyrrolidine hydrobromide

Synonym(s):

2-(bromomethyl)pyrrolidine hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H11N1Br2
CAS Number:
Molecular Weight:
244.96
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H10BrN.BrH/c6-4-5-2-1-3-7-5;/h5,7H,1-4H2;1H

Inchi Key

XBSPZBZXDDWESK-UHFFFAOYSA-N

Purity

0.95

Storage temp.

Freezer/Argon

Country of Origin

US



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library