Skip to Content
Merck

Skip To

MATH9C56E0E2

Matrix Scientific

(2R,3S,4S,5S,6R)-6-(((3E,5E,8S,9E,11S,12R,13E,15E,18S)-12-(((2R,3S,4R,5S)-3,4-Dihydroxy-5-(isobutyryloxy)-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-11-ethyl-8-hydroxy-18-((R)-1-hydroxyethyl)-9,13,15-trimethyl-2-oxooxacyclooctadeca-3,5,9,13,15-pentaen-3-yl)methoxy)-4-hydroxy-5-methoxy-2-methyltetrahydro-2H-pyran-3-yl 3,5-dichloro-2-ethyl-4,6-dihydroxybenzoate

Synonym(s):

(2R,3S,4S,5S,6R)-6-(((3Z,5Z,8S,9Z,11S,12R,13Z,15Z,18S)-12-(((2R,3S,4R,5S)-3,4-dihydroxy-5-(isobutyryloxy)-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-11-ethyl-8-hydroxy-18-((R)-1-hydroxyethyl)-9,13,15-trimethyl-2-oxooxacyclooctadeca-3,5,9,13,15-pentaen-3-yl)methoxy)-4-hydroxy-5-methoxy-2-methyltetrahydro-2H-pyran-3-yl 3,5-dichloro-2-ethyl-4,6-dihydroxybenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C52H74Cl2O18/c1-13-30-22-26(6)33(56)18-16-15-17-31(23-66-51-45(65-12)42(61)44(29(9)67-51)69-49(64)35-32(14-2)36(53)39(58)37(54)38(35)57)48(63)68-34(28(8)55)20-19-25(5)21-27(7)43(30)70-50-41(60)40(59)46(52(10,11)72-50)71-47(62)24(3)4/h15-17,19,21-22,24,28-30,33-34,40-46,50-51,55-61H,13-14,18,20,23H2,1-12H3/b16-15-,25-19-,26-22-,27-21-,31-17-/t28-,29-,30+,33+,34+,40-,41+,42+,43+,44-,45+,46+,50-,51-/m1/s1

Inchi Key

ZVGNESXIJDCBKN-VUCVDCOISA-N

Purity

0.98

Storage temp.

Refrigerate

Country of Origin

US

Boiling Point

1046.4?A65.0

Melting Point

161






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library