Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41116116
Quality Segment
form
liquid
specific analyte(s)
lead
technique(s)
photometry: suitable
pH
4 (20 °C in H2O)
density
1.00 g/cm3 at 20 °C
application(s)
aquaculture
boiler and cooling water
cannabis testing
disposal drainage water
drinking water
electroplating
environmental
mineral water
wastewater
storage temp.
15-25°C
General description
This Spectroquant® Lead Standard Solution is a high-quality laboratory solution used for internal quality control and method-controlling purposes.
These Spectroquant® standard solutions perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.
Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.
These Spectroquant® standard solutions perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.
Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.
Application
Lead Standard Solution can be used to check the photometric measurement system (reagents, measurement device, handling) and the mode of working.
Features and Benefits
- Manufactured and tested in DIN EN ISO 9001:2008 accredited laboratory
- The solution is pre-diluted to a concentration that falls into the range of our Spectroquant® test kits. This eliminates the need for dilution prior to analysis and hence avoids a common source of error
- Ready-to-use diluted reference materials (RMs) along with expanded measurement uncertainty ensure absolute precision
- Accompanied with a certificate of analysis (COA) detailing the certified values, including uncertainty and expiry date
- Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST) or to United States Pharmacopeia (USP)
- Reliable and reproducible results
Analysis Note
Concentration: 0.0500 ± 0.0040 mg/L Pb in HNO3
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Lead Standard Solution
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
10099-74-8
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.