Skip to Content
Merck

Skip To

144232

Replacement Impeller Drum

for use with RCS® Standard

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.74
UNSPSC Code:
41122409


packaging

pkg of 1 kit

application(s)

air monitoring
food and beverages

compatibility

for use with RCS® Standard

shipped in

ambient

storage temp.

room temp

General description

940511
Spare part for the sampling head of the RCS Standard.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters