Skip to Content
Merck

202M-TEW

Anti-Actin, Smooth Muscle (1A4) Mouse Monoclonal Antibody

1mg/mL carrier free

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1A4, monoclonal
Application:
—
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

clone

1A4, monoclonal

description

Purified mouse monoclonal antibody in buffer containing PBS without preservatives

species reactivity

human

packaging

vial of

concentration

1 mg/mL

shipped in

wet ice

storage temp.

2-8°C

Analysis Note

Research Use Only (RUO)


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library