Skip to Content
Merck

Skip To

5249

Modified Scholten Broth

bottle filling volume 25 mL, ready-to-use, box of 10 × 25 mL, for the growth of host-bacteria in the enumeration of somatic coliphages in samples of any type of water, sediment and sludge

Synonym(s):

MS media, MSB, Modified Scholten′s Media, Scholten′s Broth Modified

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 x 125 mL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

1 460,00 CZK

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74
eCl@ss:
32039090
Bottle capacity:
125 mL
Bottle filling volume:
25 mL
Feature:
ready-to-use
Agency:
ISO 10705-2, ISO 2000
Ph:
7.4 ± 0.2 (25 °C)

1 460,00 CZK


Available to ship TODAYDetails


​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 10705-2, ISO 2000

form

gelatinous solid

feature

ready-to-use

packaging

box of 10 × 25 mL

technique(s)

microbiological culture: suitable

bottle capacity

125 mL

bottle filling volume

25 mL

color

yellow

pH

7.4 ± 0.2 (25 °C)

application(s)

food and beverages
water monitoring

compatibility

for use with VIRAPREP Kit (AR00206)

storage temp.

2-8°C

suitability

selective for bacteria

General description

Modified Scholten Broth  (MSB) is a selective and differential microbial culture media. This ready-to-use media comes in a semi-solid form as described in ISO 10705-2: 2000.
 
Modified Scholten Broth  (MSB) contains a higher amount of salt in the composition which inhibits the growth of non-halophilic (non salt-loving) microbes. Mannitol acts as the energy source, while phenol red serves as a pH indicator, changing color to confirm the presence of target microorganisms.

Application

Modified Scholten Broth (MSB) is used to enumerate somatic coliphages from water, sediment and sludge samples.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
524710X200ML524810X100ML17171
description

bottle filling volume 25 mL, ready-to-use, box of 10 × 25 mL, for the growth of host-bacteria in the enumeration of somatic coliphages in samples of any type of water, sediment and sludge

description

ready-to-use, box of 10 × 200 ± 5 mL, selective for bacteria

description

ready-to-use, box of 10 × 125 ± 3 mL, selective for bacteria

description

NutriSelect® Plus, powder, nonselective and differential for Escherichia coli, pack of 500 g, suitable for microbiology

suitability

selective for bacteria

suitability

selective for bacteria

suitability

-

suitability

-

feature

ready-to-use

feature

ready-to-use

feature

ready-to-use

feature

-

application(s)

food and beverages
water monitoring

application(s)

food and beverages
water monitoring

application(s)

food and beverages
water monitoring

application(s)

microbiology, agriculture
cannabis testing
environmental
food and beverages
water monitoring

form

gelatinous solid

form

box (Semi Solid Media Filled)

form

-

form

-

bottle capacity

125 mL

bottle capacity

250 mL

bottle capacity

-

bottle capacity

-

bottle filling volume

25 mL

bottle filling volume

-

bottle filling volume

-

bottle filling volume

-


Storage Class

12 - Non Combustible Liquids

WGK

nwg



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library