Skip to Content
Merck

Skip To

69002

Perfect DNA Markers, 0.5-12 kbp - Novagen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
4 080,00 CZK

About This Item

NACRES:
NA.31
UNSPSC Code:
12352200

4 080,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
692787053870539
description

-

description

-

description

-

description

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

Perfect DNA Markers 0.5-12 kbp Novagen

-

-

-


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library