Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
8 mL | Estimated to ship on11 November 2026fromAreál Kühne+Nagel spol. s r.o. | 12 500,00 CZK |
About This Item
NACRES:
NA.32
UNSPSC Code:
12352202
Quality Segment
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description Cytiva 38152103011150, 1 μm avg. part. size, suspension (1% Solids) | description 70 μM in H2O |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 200 |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



