Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 reactions | Estimated to ship on20 November 2026fromAreál Kühne+Nagel spol. s r.o. | 6 680,00 CZK |
About This Item
General description
AccepTor Vectors are supplied in a ready-to-ligate format. Simply mix the vector with your PCR product, add Clonables 2X Ligation Premix (Cat No. 70573), and transform into NovaBlue Singles Competent Cells (Cat. No. 70181). The entire procedure, from PCR product to plating transformants, can be performed in as little as 40 minutes with minimal pipetting.
AccepTor Vector Kits are available in an introductory 10-reaction size as well as 20- and 40-reaction configurations. The AccepTor Vectors are also available separately without the ligation and transformation components. Also see pSTBlue-1 AccepTor Vector Giga Kit (Cat. No. 71228) for information on using higher-efficiency competent cells.
Two different vectors, pSTBlue-1 and pETBlue-1, are available as AccepTor Vector kits. Each is carefully prepared and tested for optimal cloning efficiency and provides easy visualization of recombinants by blue/white screening using lacZ α-complementation. The pSTBlue-1 is a general purpose vector with dual opposed T7 and SP6 promoters, both with amp and kan resistance, and an array of flanking restriction sites. The pETBlue-1 vector is a novel plasmid specifically designed to enable high-level T7 RNA polymerase-driven expression of target genes, while providing the convenience of a blue/white screening method. Screening is independent of expression because the associated T7lac expression promoter is in an opposed orientation relative to the E. coli promoter. pETBlue-1 facilitates the expression of native unfused proteins and allows for convenient subcloning of target genes already fused to existing detection and purification tags. An EcoR V cloning site is appropriately spaced downstream of an E. coli ribosome binding site. Inserts must encode an ATG start codon at their 5′ end if expression is desired. The sequence is numbered from the first base of the T7 promoter sequence.
Features and Benefits
- No restriction enzymes or special primers
- Direct ligation of PCR product with vector
- Compatible with polymerases that leave single 3´-dA overhangs
- Blue/white screening with pSTBlue-1 or pETBlue-1 vectors
- Simple protocol takes as little as 40 minutes from PCR product to plating transformants
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| storage temp. −20°C | storage temp. −70°C | storage temp. −70°C | storage temp. −70°C |
| shipped in dry ice, wet ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| storage condition OK to freeze | storage condition OK to freeze | storage condition OK to freeze | storage condition OK to freeze |
| Quality Level 200 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
AccepTor Vector for direct ligation | - | Novagen pETBlue-1 vector cloning kit, pETBlue-1 cloning vector kit Sigma-Aldrich, recombinant Taq polymerase pETBlue-1 kit, laboratory cloning expression reagents, non-proofreading DNA polymerase cloning kit, pETBlue-1 vector for gene cloning, AccepTor Vector Kit for transformation, 3′ dA overhang cloning vector kit, Sigma-Aldrich cloning and expression kit, dU to dT conversion vector kit, PCR cloning pETBlue-1 vector kit, KOD XL polymerase AccepTor Vector Kit, pETBlue-1 AccepTor Vector Kit, direct ligation pETBlue-1 AccepTor Vector | - |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


