Skip to Content
Merck

Skip To

71087

KOD XL DNA Polymerase

High fidelity DNA polymerase designed for accurate PCR amplification of long strand and GC- rich DNA templates. Excellent for incorporation of deriviatized dNTPs.

Synonym(s):

High-fidelity DNA polymerase, KOD polymerase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 units

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

12 900,00 CZK
1250 units

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

77 100,00 CZK

About This Item

UNSPSC Code:
41106314
NACRES:
NA.55

12 900,00 CZK


Available to ship TODAYDetails


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
710857108671842
description

High fidelity DNA polymerase designed for accurate PCR amplification of long strand and GC- rich DNA templates. Excellent for incorporation of deriviatized dNTPs.

description

High fidelity DNA polymerase designed for accurate PCR amplification of DNA templates for general cloning and cDNA amplification applications.

description

High fidelity DNA polymerase designed for accurate PCR amplification of long strand and GC- rich DNA templates for cloning and cDNA amplification applications.

description

ready-to-use solution, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., suitable for PCR

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters