Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.79
UNSPSC Code:
41121521
eCl@ss:
32039090
Quality Segment
storage condition
room temperature
technique(s)
PCR: suitable
application(s)
food and beverages
compatibility
BioControl GDS
Application
PCR
Other Notes
One UK Power Cord
Legal Information
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.