Skip to Content
Merck

Skip To

902822

Rapid-action nipple with tube connection 6x4 mm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 unit
Please contact Customer Service for Availability
11 000,00 CZK

About This Item

NACRES:
NB.51
UNSPSC Code:
41121815
eCl@ss:
32039090

11 000,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

ambient

storage temp.

room temp

General description

Our withdrawal and safety components are easily interconnectable thanks to a wide selection of reducers, adapters and couplings. Due to the wide variety, many different customized installations are possible – everything for your individual requirements.
This rapid-action connector is made of stainless steel with a special high quality sealing for maximum protection of our solvents quality. It is equipped with nipple (male) Type 21 and tube connector 6x4 mm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
928891902267902265
description

-

description

-

description

3/8 in. (product side)

description

stainless steel connector, female connector for Type 21 coupling, for use with 8 x 6mm gas feed tube

storage temp.

room temp

storage temp.

room temp

storage temp.

no temp limit

storage temp.

no temp limit

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters