Skip to Content
Merck

9Q0418

Anti-buprenorphine(1mg) KC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
monoclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

clone

monoclonal

species reactivity

mouse, all

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable

isotype

IgG1

General description

Buprenorphine is an opoid that is frequently used to treat addiction to other opoids such as heroin or oxycodone

Preparation Note

2-8°C. For long-term storage, freeze at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library