Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
monoclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
-
Pricing and availability is not currently available.
antibody form
purified antibody
Quality Segment
clone
monoclonal
species reactivity
all, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable
isotype
IgG1
General description
Barbiturates are a class of controlled drug that act as depressants of the central nervous system. Both phycologically and physically addictive barbiturates are often abused for recreational use and are frequently included in basic drug screens
Application
Suggested antibody pairs MABX4081-1KC and MABX4082-1KC
Preparation Note
2-8°C. For long-term storage, freeze at -20°C.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Barbituate (1mg) KC
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.