Skip to Content
Merck

Skip To

AABPC5000

MCE Membrane Filter, 0.8 μm Pore Size

MF-Millipore™, filter diam. 50 mm, hydrophilic, black

Sustainability features

Synonym(s):

MF-Millipore™ Membrane Filter, 0.8 µm pore size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 ea

Estimated to ship on09 November 2026fromAreál Kühne+Nagel spol. s r.o.

5 470,00 CZK

About This Item

UNSPSC Code:
40161507
NACRES:
NB.22
eCl@ss:
32031602
Pore size:
0.8 μm pore size
Material:
black filter, mixed cellulose esters (MCE) membrane, plain filter
Feature:
hydrophilic
Diam.:
50 mm

5 470,00 CZK


Estimated to ship on09 November 2026Details


​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

black filter, mixed cellulose esters (MCE) membrane, plain filter

description

50 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, black, 100 discs

feature

hydrophilic

manufacturer/tradename

MF-Millipore™, Millipore

sustainability

Sustainability features

diam.

50 mm

filter diam.

50 mm

thickness

120 μm

color

black

matrix

MF-Millipore™

pore size

0.8 μm pore size

greener alternative category

shipped in

ambient

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.



MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.



Features & Benefits:

- Versatile filter for biological and environmental monitoring applications

- Available in a range of pore sizes, colored black or white, with or without a gridded surface

- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Fluorescent assays
  • Particle monitoring
  • Air monitoring
  • Microplastics

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AABG04700AAWG01300PHWP04700
description

MF-Millipore™, filter diam. 50 mm, hydrophilic, black

description

MF-Millipore™, filter diam. 47 mm, hydrophilic, black, gridded

description

MF-Millipore™, filter diam. 13 mm, hydrophilic, white, gridded

description

MF-Millipore™, filter diam. 47 mm, hydrophilic

diam.

50 mm

diam.

47 mm

diam.

13 mm

diam.

47 mm

manufacturer/tradename

Millipore, MF-Millipore™

manufacturer/tradename

MF-Millipore™

manufacturer/tradename

Millipore, MF-Millipore™

manufacturer/tradename

Millipore, MF-Millipore™

pore size

0.8 μm pore size

pore size

0.8 μm pore size, 82 % porosity

pore size

0.8 μm pore size, 82 % porosity

pore size

77 % porosity, 0.3 μm pore size

material

mixed cellulose esters (MCE) membrane, black filter, plain filter

material

mixed cellulose esters (MCE) membrane, gridded , black filter

material

mixed cellulose esters (MCE) membrane, white filter, gridded

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

description

50 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, black, 100 discs

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, black, gridded, 100 discs

description

13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 100 discs

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs


Still not finding the right product?


Signal Word

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Irrit. 2 - Flam. Sol. 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9036-19-5

CAS No.

9036-19-5

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters