Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 μg | Please contact Customer Service for Availability | 14 900,00 CZK |
About This Item
NACRES:
NA.41
UNSPSC Code:
51111800
Clone:
-
Species reactivity:
-
Application:
—
Citations:
-
Quality Segment
1 of 1
This Item | |
|---|---|
| description - | description - |
| Quality Level 300 | Quality Level 300 |
Rb Olfactory Receptor Family 2 antibody Sigma-Aldrich | - |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

