Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship on04 November 2026fromAreál Kühne+Nagel spol. s r.o. | 11 500,00 CZK |
About This Item
conjugate
unconjugated
Quality Segment
antibody form
affinity purified immunoglobulin
clone
polyclonal
purified by
(Antigen Affinity Purified)
species reactivity
human, rabbit
concentration
(Please refer to lot specific datasheet.)
technique(s)
western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
Application
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis (WB): 1 μg/mL of this antibody detected IDH1 in HepG2 cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description - | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description affinity isolated antibody, buffered aqueous solution |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| species reactivity human, rabbit | species reactivity human | species reactivity human, mouse, rat | species reactivity human, rat, mouse |
| technique(s) western blot: suitable | technique(s) immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:200-1:500 | technique(s) immunohistochemistry: 1:500-1:1000, immunoblotting: 0.04-0.4 μg/mL | technique(s) indirect ELISA: suitable, western blot: suitable |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



