Skip to Content
Merck

Skip To

ABC1484

Anti-Bcl-B Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on01 December 2026fromAreál Kühne+Nagel spol. s r.o.

11 500,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-

11 500,00 CZK


Estimated to ship on01 December 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Segment

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

(Antigen Affinity Purified)

species reactivity

rabbit, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable, western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

Bcl-B antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human Bcl-B.
~40 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Application

Epitope: Near N-terminus

Physical form

Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis (WB): 1 μg/mL of this antibody detected Bcl-B in Jurkat cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B3170SAB5500015SAB4500003
description

-

description

clone Bcl-2-100, ascites fluid

description

clone SP155, recombinant, expressed in proprietary host, affinity isolated antibody

description

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

ascites fluid

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

clone

polyclonal

clone

Bcl-2-100, monoclonal

clone

SP155, monoclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

rabbit, human

species reactivity

human

species reactivity

human (tested)

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, electron microscopy: suitable, immunoprecipitation (IP): suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: 1:1,000 using human HeLa cell extract, immunohistochemistry (frozen sections): suitable, microarray: suitable

technique(s)

flow cytometry: 1:100, immunohistochemistry: 1:100

technique(s)

western blot: 1:500-1:1000, immunohistochemistry: 1:50-1:100, ELISA: 1:10000, immunofluorescence: 1:100-1:500

UniProt accession no.

Q9HD36

UniProt accession no.

P10415

UniProt accession no.

P41182

UniProt accession no.

P10415


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library