Skip to Content
Merck

Skip To

ABC343

Anti-Caspase-10 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Casp-10, Caspase-10

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
12 500,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

12 500,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP10(843)

General description

Caspase-10 is a member of the peptidase C14A family and is involved in the activation cascade of caspases responsible for apoptosis execution. Caspase-10 has two death effector domains (DEDs) that bind to the DED in the adapter molecule FADD and recruits both TNFR1 and CD95 to form complexes with these receptors. Caspase-10 is expressed in many tissues and cell lines with the lowest expression found in brain, kidney, prostate, testis, and colon. Mutations in Caspase-10 have been associated with autoimmune lymphoproliferative syndrome type 2A and gastric cancer.
~59 kDa observed. Uniprot describes 6 isoforms ranging from ~31 kDa to ~59 kDa Uncharacterized bands may appear in some lysates.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human Caspase-10.

Application

Detect Caspase 10 using this rabbit polyclonal antibody, Anti-Caspase-10 Antibody validated for use in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody recognizes the C-terminus of Caspase-10.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa, Jurkat, A431, and K562 whole cell lysates
Evaluated by Western Blotting in HeLa, Jurkat, A431, and K562 whole cell lysates.

Western Blotting Analysis: 1 µg/mL of this antibody detected Caspase-10 in 15 µg of HeLa, Jurkat, A431, and K562 whole cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0000843M1HPA017059C7611
description

from rabbit, purified by affinity chromatography

description

clone 1D8, purified immunoglobulin, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

clone 14F7, purified immunoglobulin, buffered aqueous solution

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

UniProt accession no.

Q92851

UniProt accession no.

Q92851

UniProt accession no.

Q92851

UniProt accession no.

Q920D5

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

rat, mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


WGK

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters