Skip to Content
Merck

Skip To

ABC458

Anti-Thymidylate Kinase/TMPK Antibody

from rabbit

Synonym(s):

Thymidylate kinase, dTMP kinase

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
1


biological source

rabbit

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TK1(7083)

General description

TMPK, also known as dTMP kinase, and encoded by the gene DTYMK/CDC8/TMPK/TYMK, is a key enzyme for pyrimidine synthesis that catalyzes the phosphorylation of thymidine 5′-monophosphate (dTMP) to form thymidine 5′-diphosphate (dTDP). TMPK has important functions in cell proliferation and DNA replication, and the synthesis of dTDP is unique because it requires TMPK. All other dNDPs are directly produced by ribonucleotide reductase (RNR). Thus, TMPK is well recognized as a potential drug target for many diseases and disease agents including viruses. TMPK is currently a target of anti-HIV nucleoside therapy, and TMPK is also a validated target for antibiotic development against resistance strains of bacteria and parasites, and research demonstrates that it may also be a good target for the anticancer agent doxorubicin cancer cells. Expressed by virtually all cells it is localized to cell cytoplasm with only traces found in Golgi, mitochondria and nuclei.
~24 kDa observed

Immunogen

GST-tagged recombinant protein corresponding to human Thymidylate Kinase/TMPK.

Application

Anti-Thymidylate Kinase/TMPK Antibody is a rabbit polyclonal antibody, validated for use in western blotting & IP.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: A representative lot from an independent laboratory detected Thymidylate Kinase/TMPK in MDA-MB231, MCF-7, H184B5F5/M10 cell lysates (Hu, C. M., et al. (2012). Cancer Cell. 22(1):36-50.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunprecipitated Thymidylate Kinase/TMPK from MCF-7, H184B4F5/M10, and MCF-10A cell lysates (Ke, P. Y., et al. (2005). Genes Dev. 19(16):1920-1933.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Thymidylate Kinase/TMPK in MCF-7, H184B4F5/M10, and MCF-10A cell lysates (Ke, P. Y., et al. (2005). Genes Dev. 19(16):1920-1933.).

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Thymidylate Kinase/TMPK in 10 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters