Skip to Content
Merck

Skip To

ABC481

Anti-Cytosolic 5′-nucleotidase II Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

affinity chromatography

species reactivity

rabbit, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

KLH-conjugated linear peptide corresponding to human Cytosolic 5′-nucleotidase II near the C-terminus.
~65 kDa observed. This antibody is expected to detect both isoforms.

Application

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected Cytosolic 5′-nucleotidase II in human kidney tissue.

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Cytosolic 5′-nucleotidase II in 10 µg of A549 cell lysate.

Other Notes

Human. Predicted to react with Mouse, Rat, Feline, Canine, Bovine, and Horse based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4503371HPA021581HPA023632
description

-

description

affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

rabbit, human

species reactivity

mouse, human

species reactivity

rat, human

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

western blot: 1:500-1:1000, ELISA: 1:20000, immunofluorescence: 1:100-1:500

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:20-1:50

technique(s)

immunohistochemistry: 1:200-1:500, immunofluorescence: 0.25-2 μg/mL

UniProt accession no.

P49902

UniProt accession no.

-

UniProt accession no.

Q8TCD5

UniProt accession no.

Q8TCD5




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library