Skip to Content
Merck

Skip To

ABC498

Anti-BCL2A1 Antibody/BFL-1

serum, from rabbit

Synonym(s):

Bcl-2-related protein A1, Bcl-2-like protein 5, Bcl2-L-5, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship on19 November 2026fromAreál Kühne+Nagel spol. s r.o.

12 200,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1

12 200,00 CZK


Estimated to ship on19 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCL2A1(597)

General description

Bcl-2-related protein A1 (BCL2A1), also known as protein BFL-1, protein GRS, and Bcl-2-like protein 5 (Bcl2-L-5), is a member of the Bcl-2 family. BCL2A1/BFL-1 can interact with Bax and suppress apoptosis by inhibiting the release of cytochrome c and caspase-3 activation. BCL2A1/BFL-1 is expressed in peripheral blood, spleen, bone marrow, lung, small intestine, and testis.
~20 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Recombinant protein corresponding to human BCL2A1/BFL-1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-BCL2A1 Antibody/BFL-1 is validated for use in Western Blotting and Immunohistochemistry and Immunoprecipitation for the detection of BCL2A1/BFL-1.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected BCL2A1/BFL-1 in 0.2 µg of Jurkat cell lysate.
Western Blotting Analysis: A representative lot detected BCL2A1/BFL-1 in human lymphoma cell lysate (Yecies, D., et al. (2010). Blood. 115(16):3304-13).
Western Blotting Analysis: A representative lot detected BCL2A1/BFL-1 in chronic lymphocytic leukemia cell lysate (Natoni, A., et al. (2011). Mol Cancer Ther. 10(9):1624-1634).
Immunohistochemistry Analysis: A representative lot detected BCL2A1/BFL-1 in lymph node tissue (Vogler, M., et al. (2009). Blood. 113(18): 4403-4413).
Immunoprecipitation Analysis: A representative lot immunoprecipitated BCL2A1/BFL-1 in chronic lymphocytic leukemia cell lysate (Vogler, M., et al. (2009). Blood. 113(18): 4403-4413).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in human BFL-1 recombinant protein.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected BCL2A1/BFL-1 in 0.2 µg of human BFL-1 recombinant protein.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2900263SAB2501619PRS3873
description

serum, from rabbit

description

affinity isolated antibody, buffered aqueous solution

description

affinity isolated antibody, buffered aqueous solution

description

affinity isolated antibody, buffered aqueous solution

antibody form

serum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

goat

biological source

rabbit

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable, immunohistochemistry: suitable

technique(s)

western blot: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

-

technique(s)

-

species reactivity

human

species reactivity

-

species reactivity

bovine, canine, human, pig

species reactivity

human, mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters