Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship on26 October 2026fromAreál Kühne+Nagel spol. s r.o. | 11 900,00 CZK |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
bovine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), canine (based on 100% sequence homology), horse (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PAWR(5074)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1.0 µg/mL of this antibody detected PAR-4 in 10 µg of Y-79 cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description from rabbit, purified by affinity chromatography | description ascites fluid, clone 3G9H7, from mouse | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description - |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form ascites fluid | antibody form affinity isolated antibody | antibody form - |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| technique(s) western blot: suitable | technique(s) western blot: suitable, immunohistochemistry: suitable | technique(s) immunohistochemistry: 1:50-1:200, immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL | technique(s) western blot: 1:500 - 1:2000, immunofluorescence: 1:50 - 1:200, immunohistochemistry: 1:50 - 1:200 |
| clone polyclonal | clone 3G9H7, monoclonal | clone polyclonal | clone 10Q2G9, monoclonal |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



