Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 μL | Estimated to ship on23 November 2026fromAreál Kühne+Nagel spol. s r.o. | 11 900,00 CZK |
About This Item
conjugate
unconjugated
Quality Segment
antibody form
affinity purified immunoglobulin
clone
polyclonal
purified by
affinity chromatography
species reactivity
rabbit, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
Application
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: A 1:250 dilution of this antibody detected MATE1 in 10 µg of human kidney tissue lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description - | description affinity isolated antibody | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody | description from rabbit, purified by affinity chromatography |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| species reactivity rabbit, human | species reactivity human | species reactivity human | species reactivity human, rat |
| technique(s) western blot: suitable | technique(s) immunoblotting: suitable | technique(s) immunofluorescence: 0.25-2 μg/mL | technique(s) western blot: suitable, immunohistochemistry: suitable (paraffin) |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



