Skip to Content
Merck

Skip To

ABC830

Anti-SEC62 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Translocation protein SEC62, Translocation protein 1, TP-1, hTP-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on30 November 2026fromAreál Kühne+Nagel spol. s r.o.

11 900,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

11 900,00 CZK


Estimated to ship on30 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SEC62(7095)

General description

Translocation protein SEC62, also known as Translocation protein 1 (TP-1) or (hTP-1), and encoded by the gene name SEC62, is an integral membrane protein located in the rough endoplasmic reticulum (ER) and is part of the SEC61-SEC62-SEC63 complex. This complex is the central component of the protein translocation apparatus of the ER membrane. It is speculated that SEC61-SEC62-SEC63 may perform post-translational protein translocation into the ER and might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway. Silencing of this gene with RNAi dramatically reduced the migration and invasive potential of numerous tumor cell lines, suggesting that it may be an attractive target for therapy of various tumors.
~31 and ~43 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: C-terminus
KLH-conjugated synthetic peptide corresponding to the C-terminus of human SEC62.

Application

Anti-SEC62 Antibody detects level of SEC62 & has been published & validated for use in SEC62.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected SEC62 in mouse brain tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody is predicted to not cross-react with other SEC proteins.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 0.5 μg/mL of this antibody detected SEC62 in rat brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA014059SAB1303608SAB5700119
description

from rabbit, purified by affinity chromatography

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

IgG fraction of antiserum, buffered aqueous solution

description

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... SEC62(7095)

Gene Information

human ... SEC62(7095)

Gene Information

-

Gene Information

-

technique(s)

western blot: suitable, immunohistochemistry: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:50-1:200, immunofluorescence: 0.25-2 μg/mL

technique(s)

-

technique(s)

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


WGK

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters