Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Please contact Customer Service for Availability | 11 900,00 CZK |
About This Item
conjugate
unconjugated
Quality Segment
antibody form
affinity purified immunoglobulin
clone
polyclonal
purified by
affinity chromatography
species reactivity
rabbit, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunohistochemistry: suitable, western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
Application
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1.0 µg/mL of this antibody detected AMACR in 10 µg of human kidney tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description - | description affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, clone CL9362, purified immunoglobulin, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, clone CL9360, purified immunoglobulin, buffered aqueous glycerol solution |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone polyclonal | clone CL9362, monoclonal | clone CL9360, monoclonal |
| species reactivity rabbit, human | species reactivity human | species reactivity human | species reactivity human |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| UniProt accession no. | UniProt accession no. | UniProt accession no. - | UniProt accession no. - |
| technique(s) western blot: suitable, immunohistochemistry: suitable | technique(s) western blot: 0.04-0.4 μg/mL, immunohistochemistry: 1:2500-1:5000 | technique(s) immunohistochemistry: 1:500-1:1000, western blot: 1 μg/mL | technique(s) immunohistochemistry: 1:200-1:500, western blot: 1 μg/mL |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



