Skip to Content
Merck

Skip To

ABC95

Anti-Praf2 (JM4) Antibody

serum, from rabbit

Synonym(s):

PRA1 family protein 2, Prenylated Rab acceptor 1 family protein 2, PRAF2, JM4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship on12 November 2026fromAreál Kühne+Nagel spol. s r.o.

12 200,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

12 200,00 CZK


Estimated to ship on12 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), dog (based on 100% sequence homology), hamster (based on 100% sequence homology), guinea pig (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRAF2(11230)

General description

Prenylated Rab acceptor 1 family protein 2 (PRAF2), or JM4, is a novel 19-kDa transmembrane protein belonging to the PRA1 family. It is highly expressed in the brain, and has been shown to be enriched in synaptic vesicles. PRAF2 is also relatively abundant in the spleen, small intestine, lung, pancreas, as well as various tumors affecting the ovary, breast, and colon. Recent studies have identified PRAF2 as a potential marker for neuroblastoma. Like other PRAF1 proteins, it is localized to the golgi, endoplasmic reticulum, and vesicular structures. The precise role of PRAF2 is not yet clear, but studies suggest that it may play a role in cellular transport processes.
~18 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human Praf2 (JM4).

Application

Anti-Praf2 (JM4) Antibody is an antibody against Praf2 (JM4) for use in Western Blotting & IHC.
Immunohistochemistry Analysis: A representative lot detected Praf2 (JM4) in mouse and human cerebellum tissue and in human breast cancer tissue.
Research Category
Apoptosis & Cancer

Neuroscience
Research Sub Category
Apoptosis - Additional

Synapse & Synaptic Biology

Physical form

Rabbit polyclonal serum containing with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
SK-N-SH cell lysate
Western Blot Analysis: A 1:2,000 dilution from a representative lot detected Praf2 (JM4) in 10 µg of SK-N-SH cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA029171SAB3500375HPA015540
description

serum, from rabbit

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

affinity isolated antibody, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

serum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

UniProt accession no.

O60831

UniProt accession no.

Q9UI14

UniProt accession no.

Q12933

UniProt accession no.

-

species reactivity

mouse, human, rat

species reactivity

human

species reactivity

mouse, human, rat

species reactivity

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters