Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Estimated to ship on23 November 2026fromAreál Kühne+Nagel spol. s r.o. | 12 200,00 CZK |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PRUNE(58497)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Physical form
Preparation Note
Analysis Note
HeLa S3 cell lysate
Western Blot Analysis: A 1:5,000 dilution of this antibody detected PRUNE in 10 µg of HeLa S3 cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description from rabbit, purified by affinity chromatography | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone polyclonal | clone polyclonal | clone - |
| UniProt accession no. | UniProt accession no. - | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



