About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... MYSM1(114803)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Physical form
Preparation Note
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
NIH/3T3 cell lysate
Western Blot Analysis: 1:1,000 dilution of this antibody detected MYSM1 on 10 µg of NIH/3T3 cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description serum, from rabbit | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description from rabbit, purified by affinity chromatography |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| antibody form serum | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| UniProt accession no. | UniProt accession no. | UniProt accession no. - | UniProt accession no. |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



