Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship on23 October 2026fromAreál Kühne+Nagel spol. s r.o. | 11 500,00 CZK |
About This Item
Quality Segment
biological source
rabbit
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
technique(s)
immunocytochemistry: suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ESR1(2099)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Analysis Note
Western Blot Analysis: 2 µg/mL of this antibody detected ERR-alpha on 10 µg of HeLa cell lysate.
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description from rabbit | description Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description from rabbit, purified by affinity chromatography |
| antibody form purified antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| species reactivity human, mouse | species reactivity human | species reactivity human | species reactivity mouse |
| Gene Information human ... ESR1(2099) | Gene Information human ... ESR1(2099) | Gene Information human ... ESR1(2099) | Gene Information human ... ESRRB(2103) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



