Skip to Content
Merck

Skip To

ABS461

Anti-SHOC2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich repeat protein SHOC-2, Ras-binding protein Sur-8, SOC2, Protein soc-2 homolog, Protein sur-8 homolog

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on05 November 2026fromAreál Kühne+Nagel spol. s r.o.

11 500,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

11 500,00 CZK


Estimated to ship on05 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SHOC2(8036)

General description

The Leucine-rich repeat protein SHOC-2 (SHOC2) is the regulatory subunit of protein phosphatase 1 (PP1c) that acts as a M-Ras/MRAS effector and plays a key role in MAPK pathway activation. Upon RAS activation, SHOC2 acts as a positive modulator of the RAS-MAPK signaling via specifically allowing PP1c to dephosphorylate the ′Ser-259′ inhibitory site of RAF1 kinase and stimulate RAF1 activity. SHOC2 localizes to cytoplasm when inactive but rapidly translocates to the nucleus when activated, such as during growth factor stimulation. Thus, as expected, SHOC2 also participates in FGF receptor signaling. Molecularly, SHOC2 is a scaffold protein that brings together activated Ras (activated via G-protein coupled receptors and the interaction of GTP to GDP) and Raf, which then facilitates the transfer of the phospho-Raf phosphate to other proteins for the activation of the MAP Kinase pathway for gene activation. Defects in SHOC2 lead to a type of multi-symptomatic disorder termed the Noonan Syndrome Loss of Anagen Hair (NSLAH) that affects everything from skeletal formation to facial features and many organs including the brain, skin, muscles, and heart. EMD Millipore’s Anti-SHOC2 rabbit polyclonal antibody has been tested in western blotting and immunohistochemistry in human samples.
~65 kDa observed. Uniprot describes 2 isoforms at ~60 kDa and ~65 kDa Uncharacterized bands may appear in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to human SHOC2.

Application

Detect Leucine-rich repeat protein SHOC-2 using this rabbit polyclonal antibody, Anti-SHOC2 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected SHOC2 in human spleen tissue.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate
Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected SHOC2 in 15 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB3500051HPA009164HPA048134
description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

UniProt accession no.

Q9UQ13

UniProt accession no.

Q9UQ13

UniProt accession no.

Q9UQ13

UniProt accession no.

-

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


WGK

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters