Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship on06 November 2026fromAreál Kühne+Nagel spol. s r.o. | 11 500,00 CZK |
About This Item
conjugate
unconjugated
Quality Segment
antibody form
affinity purified immunoglobulin
clone
polyclonal
purified by
(Antigen Affinity Purified)
species reactivity
rabbit, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
immunocytochemistry: suitable, western blot: suitable
UniProt accession no.
target post-translational modification
unmodified
General description
Application
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected IKK beta in Jurkat whole cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description affinity isolated antibody | description affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| species reactivity rabbit, human | species reactivity mouse, rat, human | species reactivity human, mouse, rat | species reactivity human, mouse, rat |
| antibody form affinity purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| technique(s) immunocytochemistry: suitable, western blot: suitable | technique(s) immunohistochemistry: 1:50-1:200, immunofluorescence: 1:50-1:200, western blot: 1:500-1:2000 | technique(s) western blot: 1:500-1:1000, immunohistochemistry: 1:50-1:100, ELISA: 1:1000 | technique(s) ELISA: 1:5000, western blot: 1:500-1:1000, immunohistochemistry: 1:50-1:100 |
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



