Skip to Content
Merck

Skip To

DP01

Anti-SV40 T Antigen (Ab-1) Mouse mAb (PAb419)

This Anti-SV40 T Antigen (Ab-1) Mouse mAb (PAb419) is validated for use in Immunoblotting

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
12 200,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Species reactivity:
(SV40 infected cells)
Species reactivity:
-
Application:
—
Citations:
-

12 200,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

form

liquid

species reactivity

(SV40 infected cells)

storage condition

do not freeze

dilution

(Immunoblotting
Immunofluorescence (1-5 µg/mL)
Immunoprecipitation (1-2 µg/sample))

isotype

IgG2a

storage temp.

2-8°C

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells . Recognizes the ~94 kDa SV40 large T antigen and the ~20.5 kDa SV40 small T antigen.

Immunogen

the N-terminal domain of SV40 large and small T-antigen

Application

Immunoblotting Immunofluorescence (1-5 µg/mL)Immunoprecipitation (1-2 µg/sample)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DP02MABF121OP13
description

This Anti-SV40 T Antigen (Ab-1) Mouse mAb (PAb419) is validated for use in Immunoblotting

description

liquid, clone PAb416, Calbiochem®

description

clone PAb416, from mouse

description

liquid, clone NCM II 100, Calbiochem®

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

(SV40 infected cells)

species reactivity

SV40-infected cells

species reactivity

SV40-infected cells

species reactivity

human, mouse

form

liquid

form

liquid

form

-

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

isotype

IgG2a

isotype

IgG2a

isotype

IgG2aκ

isotype

IgG1

storage condition

do not freeze

storage condition

do not freeze

storage condition

-

storage condition

OK to freeze, avoid repeated freeze/thaw cycles


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters