Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Estimated to ship on19 November 2026fromAreál Kühne+Nagel spol. s r.o. | 9 620,00 CZK |
About This Item
NACRES:
NA.76
UNSPSC Code:
12352200
eCl@ss:
32160703
9 620,00 CZK
Estimated to ship on19 November 2026Details
Quality Segment
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description sufficient for 200 colorimetric or fluorometric tests |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
Signal Word
Danger
Target Organs
Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 3
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.







